Oxyresveratrol
Synonym(s):
(E)-2,3′,4,5′-Stilbenetetrol, 2,3′,4,5′-Tetrahydroxy-trans-stilbene, 4-[(1E)-2-(3,5-Dihydroxyphenyl)ethenyl]-1,3-benzenediol
Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing