
Levonordefrin

Synonym(s):
(−)-3,4-Dihydroxynorephedrine, α-(1-Aminoethyl)-3,4-dihydroxybenzyl alcohol, (−)-α-Methylnorepinephrine, 4-(2-Amino-1-hydroxypropyl)-1,2-benzenediol, L-4-(2-Amino-1-hydroxypropyl)-1,2-benzenediol, Levonordefrin
Linear Formula:
(HO)2C6H3CH(OH)CH(CH3)NH2
CAS Number:
Molecular Weight:
183.20
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing