
Isoliquiritigenin

Synonym(s):
(E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one, 4,2′,4′-Trihydroxychalcone
Empirical Formula (Hill Notation):
C15H12O4
CAS Number:
Molecular Weight:
256.25
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing