
Felbamate Related Compound A

Synonym(s):
2-Phenyl-1,3-propanediol monocarbamate, 3-Hydroxy-2-phenylpropyl carbamate
Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing