
Cocaethylene-D8 solution

Empirical Formula (Hill Notation):
C18H15D8NO4
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing