Skip to Content
Merck
cis-11,14-Eicosadienoic acid methyl ester ≥98% (GC)

cis-11,14-Eicosadienoic acid methyl ester

cis-11,14-Eicosadienoic acid methyl ester ≥98% (GC)
Empirical Formula (Hill Notation):
C21H38O2
CAS Number:
Molecular Weight:
322.53

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing