
Chlormadinone acetate impurity G

Synonym(s):
17α-Hydroxyprogesterone 17-acetate, 17α-Acetoxy-4-pregnene-3,20-dione, 17α-Acetoxyprogesterone, 17α-Hydroxy-4-pregnene-3,20-dione 17-acetate, 4-Pregnen-17α-ol-3,20-dione acetate
Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing