
Berbamine dihydrochloride

Synonym(s):
berbenine
Empirical Formula (Hill Notation):
C37H40N2O6 ·2HCl
CAS Number:
Molecular Weight:
681.65
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing