
Anti-CYBB antibody produced in rabbit

Synonym(s):
Anti-CGD, Anti-GP91-PHOX, Anti-NOX2, Anti-Cytochrome b-245, β polypeptide (chronic granulomatous disease), Anti-GP91-1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing