
(1-Methyl-4-piperidinyl)methanamine

Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing