
12-Mercaptododecanoic acid

Synonym(s):
MDA
Empirical Formula (Hill Notation):
C12H24O2S
CAS Number:
Molecular Weight:
232.38
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing