Skip to Content
Merck
All Photos(1)

Key Documents

8.07061

Sigma-Aldrich

1,5-Pentanediol

for synthesis

Synonym(s):

1,5-Pentanediol, Pentamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O2
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-854-4
NACRES:
NA.22

vapor pressure

>0.01 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

330 °C

potency

10000 mg/kg LD50, oral (Rat)
>19800 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-13.1 % (v/v)

pH

7.5 ( in H2O)

bp

240-242 °C/1013 hPa

mp

-16 °C

transition temp

flash point 136 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H12O2/c6-4-2-1-3-5-7/h6-7H,1-5H2

InChI key

ALQSHHUCVQOPAS-UHFFFAOYSA-N

Application

  • Biodegradable Polymers for Sustainable Packaging: A study on biobased multiblock copolymers incorporating 1,5-Pentanediol for sustainable food packaging explores the use of poly(l-lactic acid) and 2,5-furandicarboxylic acid, offering insights into composting kinetics and mechanisms (Bianchi et al., 2023).
  • High-Temperature Battery Applications: Research on working aqueous Zn metal batteries at 100°C examined the role of 1,5-Pentanediol in stabilizing electrolyte solutions, highlighting its utility in high-performance energy storage systems (Wang et al., 2022).
  • Biopolymer Synthesis for Environmental Sustainability: Engineered Halomonas bluephagenesis synthesizes diverse α,ψ-diol-derived polyhydroxyalkanoates, utilizing 1,5-Pentanediol, to create plastics with improved biodegradability and performance in environmental applications (Yan et al., 2022).
  • Advanced Materials with Barrier Properties: A study on renewable furfural-based polyesters containing 1,5-Pentanediol reports high oxygen barrier properties, suitable for packaging applications that require extended shelf life and sustainability (Ahmed et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.988 - 0.991
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

287.6 °F - closed cup

Flash Point(C)

142 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service