HPLC Analysis of N-(1-Phenylethyl)maleimide Enantiomers on Astec® CHIROBIOTIC® R
CONDITIONS
column
CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 μm particles (13024AST)
mobile phase
[A] 0.1% triethylamine acetate, pH 4.1; [B] methanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 254 nm
injection
3 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC