HPLC Analysis of Fluoxetine Enantiomers on Narrowbore Astec® CHIROBIOTIC® V2
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V2, 10 cm x 2.1 mm I.D., 5 μm particles (15018AST)
mobile phase
13 mM ammonium acetate in methanol
flow rate
0.2 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC