Merck
  • Facile synthesis, structure, biocompatibility and antimicrobial property of gold nanoparticle composites from cellulose and keratin.
Facile synthesis, structure, biocompatibility and antimicrobial property of gold nanoparticle composites from cellulose and keratin.
Journal of colloid and interface science (2017-09-28)
Chieu D Tran, Franja Prosenc, Mladen Franko
ABSTRACT
A novel, one-pot method was developed to synthesize gold nanoparticle composite from cellulose (CEL), wool keratin (KER) and chloroauric acid. Two ionic liquids, butylmethylimmidazolium chloride and ethylmethylimmidazolium bis(trifluoromethylsulfonyl)imide were used to dissolve CEL, KER and HAuCl
Product Description
Millipore
MRSA Selective Supplement, suitable for microbiology
Sigma-Aldrich
2-Ethylimidazole, 98%