Merck
  • Rapid and sensitive detection of free fatty acids in edible oils based on chemical derivatization coupled with electrospray ionization tandem mass spectrometry.
Rapid and sensitive detection of free fatty acids in edible oils based on chemical derivatization coupled with electrospray ionization tandem mass spectrometry.
Food chemistry (2017-10-19)
Ming Liu, Fang Wei, Xin Lv, Xu-Yan Dong, Hong Chen
ABSTRACT
In this study, a strategy based on chemical derivatization coupled with electrospray ionizationtandem quadrupole mass spectrometry (ESI-MS/MS) for rapid and sensitive detection of FFAs in edible oils was developed. A derivative reagent (N,N-diethyl-1,2-ethanediamine, DEEA) was employed to selectively label carboxyl groups of FFAs to form an amino compound with a tertiary amino group. The DEEA derivative products could lose a characteristic neutral loss fragment of 73Da in collision-induced dissociation (CID), which enabled to discriminate and analyze the DEEA derived FFAs with neutral loss scan (NLS 73Da)under the positive ion mode of mass spectrometry. The assay was linear over the concentration range 0.5-200nmol/L with satisfactory correlation coefficients (R
Sigma-Aldrich
2-Chloro-1-methylpyridinium iodide, 97%