Merck
  • Deferiprone and Gallium-Protoporphyrin Have the Capacity to Potentiate the Activity of Antibiotics in
Deferiprone and Gallium-Protoporphyrin Have the Capacity to Potentiate the Activity of Antibiotics in
Frontiers in cellular and infection microbiology (2017-07-12)
Katharina Richter, Nicky Thomas, Guimin Zhang, Clive A Prestidge, Tom Coenye, Peter-John Wormald, Sarah Vreugde
ABSTRACT
Small colony variants (SCVs) of bacteria like
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Sigma-Aldrich
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, 98%