Merck
  • Interaction of plant essential oil terpenoids with the southern cattle tick tyramine receptor: A potential biopesticide target.
Interaction of plant essential oil terpenoids with the southern cattle tick tyramine receptor: A potential biopesticide target.
Chemico-biological interactions (2016-12-18)
Aaron D Gross, Kevin B Temeyer, Tim A Day, Adalberto A Pérez de León, Michael J Kimber, Joel R Coats
ABSTRACT
An outbreak of the southern cattle tick, Rhipicephalus (Boophilus) microplus, (Canestrini), in the United States would have devastating consequences on the cattle industry. Tick populations have developed resistance to current acaricides, highlighting the need to identify new biochemical targets along with new chemistry. Furthermore, acaricide resistance could further hamper control of tick populations during an outbreak. Botanically-based compounds may provide a safe alternative for efficacious control of the southern cattle tick. We have developed a heterologous expression system that stably expresses the cattle tick's tyramine receptor with a G-protein chimera, producing a system that is amenable to high-throughput screening. Screening an in-house terpenoid library, at two screening concentrations (10 μM and 100 μM), has identified four terpenoids (piperonyl alcohol, 1,4-cineole, carvacrol and isoeugenol) that we believe are positive modulators of the southern cattle tick's tyramine receptor.
Sigma-Aldrich
Piperonyl alcohol, 98%
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Citronellic acid, 98%