Merck
  • Crystal Structures of a Series of Complexes Produced by Reaction of Copper(I) Cyanide with Diamines.
Crystal Structures of a Series of Complexes Produced by Reaction of Copper(I) Cyanide with Diamines.
Inorganic chemistry (2001-10-24)
Fred B. Stocker, Teodora P. Staeva, Chad M. Rienstra, Doyle Britton
ABSTRACT
A new synthetic procedure developed recently in our laboratories has made possible the synthesis of variety of new complexes of CuCN with diamines. Synthesis was effected by adding the ligand to a solution of CuCN in aqueous sodium thiosulfate. This procedure also provides an alternative pathway to a novel diamine complex reported by us previously, (CuCN)(3)(HMTA)(2) (1) (where HMTA = hexamethylenetetramine). The other diamine ligands used were 1,4-diazabicyclo[2.2.2]octane (dabco), 1,4-dimethylpiperazine (dmpip), piperazine (pip), 1,4-butanediamine (butda), N,N,N',N'-tetramethylethylenediamine (tetmen), and N-phenylpiperazine (phpip). Complex2, Cu(2)(CN)(3)(dabco-H), crystallizes in the hexagonal space group P6(3) with unit cell dimensions a = 8.174(3) Å, c = 8.083(4) Å, and Z = 2. Complex 3, (CuCN)(2)(dmpip), crystallizes in the monoclinic space group C2/m with unit cell dimensions a = 8.812(3) Å, b = 9.631(2) Å, c = 7.266(3) Å, beta = 113.40(3) degrees, and Z = 2. Complex 4, (CuCN)(2)(pip), crystallizes in the monoclinic space group C2/c with unit cell dimensions a = 9.439(3) Å, b = 10.561(2) Å, c = 8.870(3) Å, beta = 98.32(3) degrees, and Z = 4. Complex 5, Cu(2)(CN)(3)(pip-H), crystallizes in the monoclinic space group C2/c with unit cell dimensions a = 20.573(9) Å, b = 8.354(2) Å, c = 15.989(6) Å, beta = 133.70(3) degrees, and Z = 8. Complex 6, (CuCN)(2)(butda), crystallizes in the monoclinic space group P2(1)/c with unit cell dimensions a = 10.456(2) Å, b = 5.550(1) Å, c = 8.669(3) Å, beta = 106.80(2) degrees, and Z = 2. Complex 7, (CuCN)(2)(tetmen), crystallizes in the orthorhombic space group Cmc2(1) with unit cell dimensions a = 11.889(4) Å, b = 33.380(8) Å, c = 9.012(3) Å, and Z = 12. Complex 8, (CuCN)(phpip), crystallizes in the monoclinic space group P2(1)/cwith unit cell dimensions a = 17.8819(3) Å, b = 6.9190(1) Å, c = 8.6972(1) Å, beta = 96.720(1) degrees, and Z = 4.
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Copper(I) cyanide, 99%
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%