Merck
  • A family of drug transporters: the multidrug resistance-associated proteins.
A family of drug transporters: the multidrug resistance-associated proteins.
Journal of the National Cancer Institute (2000-08-17)
P Borst, R Evers, M Kool, J Wijnholds
ABSTRACT
The human multidrug resistance-associated protein (MRP) family currently has seven members. The ability of several of these membrane proteins to transport a wide range of anticancer drugs out of cells and their presence in many tumors make them prime suspects in unexplained cases of drug resistance, although proof that they contribute to clinical drug resistance is still lacking. Recent studies have begun to clarify the function of the MRP family members. MRPs are organic anion transporters; i.e., they transport anionic drugs, exemplified by methotrexate, and neutral drugs conjugated to acidic ligands, such as glutathione (GSH), glucuronate, or sulfate. However, MRP1, MRP2, and MRP3 can also cause resistance to neutral organic drugs that are not known to be conjugated to acidic ligands by transporting these drugs together with free GSH. MRP1 can even confer resistance to arsenite and MRP2 to cisplatin, again probably by transporting these compounds in complexes with GSH. MRP4 overexpression is associated with high-level resistance to the nucleoside analogues 9-(2-phosphonylmethoxyethyl) adenine and azidothymidine, both of which are used as anti-human immunodeficiency virus drugs. MRPs may, therefore, also have a role in resistance against nucleoside analogues used in cancer chemotherapy. Mice without Mrp1, a high-affinity leukotriene C(4) transporter, have an altered response to inflammatory stimuli but are otherwise healthy and fertile. MRP2 is the major transporter responsible for the secretion of bilirubin glucuronides into bile, and humans without MRP2 develop a mild liver disease known as the Dubin-Johnson syndrome. The physiologic functions of the other MRPs are not known. Whether long-term inhibition of MRPs in humans can be tolerated (assuming that suitable inhibitors will be found) remains to be determined.
Sigma-Aldrich
SB-MRP3-Sf9, membrane preparation for Vesicular Transport Assays, recombinant, expressed in baculovirus infected Sf9 cells
Sigma-Aldrich
SB-MRP4-HEK293, MRP4 human membrane vesicles
Sigma-Aldrich
SB-MRP3-HEK293