Merck
  • Percutaneous left atrial appendage closure vs warfarin for atrial fibrillation: a randomized clinical trial.
Percutaneous left atrial appendage closure vs warfarin for atrial fibrillation: a randomized clinical trial.
JAMA (2014-11-17)
Vivek Y Reddy, Horst Sievert, Jonathan Halperin, Shephal K Doshi, Maurice Buchbinder, Petr Neuzil, Kenneth Huber, Brian Whisenant, Saibal Kar, Vijay Swarup, Nicole Gordon, David Holmes
ABSTRACT
While effective in preventing stroke in patients with atrial fibrillation (AF), warfarin is limited by a narrow therapeutic profile, a need for lifelong coagulation monitoring, and multiple drug and diet interactions. To determine whether a local strategy of mechanical left atrial appendage (LAA) closure was noninferior to warfarin. PROTECT AF was a multicenter, randomized (2:1), unblinded, Bayesian-designed study conducted at 59 hospitals of 707 patients with nonvalvular AF and at least 1 additional stroke risk factor (CHADS2 score ≥1). Enrollment occurred between February 2005 and June 2008 and included 4-year follow-up through October 2012. Noninferiority required a posterior probability greater than 97.5% and superiority a probability of 95% or greater; the noninferiority margin was a rate ratio of 2.0 comparing event rates between treatment groups. Left atrial appendage closure with the device (n = 463) or warfarin (n = 244; target international normalized ratio, 2-3). A composite efficacy end point including stroke, systemic embolism, and cardiovascular/unexplained death, analyzed by intention-to-treat. At a mean (SD) follow-up of 3.8 (1.7) years (2621 patient-years), there were 39 events among 463 patients (8.4%) in the device group for a primary event rate of 2.3 events per 100 patient-years, compared with 34 events among 244 patients (13.9%) for a primary event rate of 3.8 events per 100 patient-years with warfarin (rate ratio, 0.60; 95% credible interval, 0.41-1.05), meeting prespecified criteria for both noninferiority (posterior probability, >99.9%) and superiority (posterior probability, 96.0%). Patients in the device group demonstrated lower rates of both cardiovascular mortality (1.0 events per 100 patient-years for the device group [17/463 patients, 3.7%] vs 2.4 events per 100 patient-years with warfarin [22/244 patients, 9.0%]; hazard ratio [HR], 0.40; 95% CI, 0.21-0.75; P = .005) and all-cause mortality (3.2 events per 100 patient-years for the device group [57/466 patients, 12.3%] vs 4.8 events per 100 patient-years with warfarin [44/244 patients, 18.0%]; HR, 0.66; 95% CI, 0.45-0.98; P = .04). After 3.8 years of follow-up among patients with nonvalvular AF at elevated risk for stroke, percutaneous LAA closure met criteria for both noninferiority and superiority, compared with warfarin, for preventing the combined outcome of stroke, systemic embolism, and cardiovascular death, as well as superiority for cardiovascular and all-cause mortality. clinicaltrials.gov Identifier: NCT00129545.
Product Description
Supelco
Warfarin, analytical standard
USP
Warfarin, United States Pharmacopeia (USP) Reference Standard
Supelco
Warfarin, PESTANAL®, analytical standard