Merck
  • Irbesartan/amlodipine: a review of its use in adult patients with essential hypertension not adequately controlled with monotherapy.
Irbesartan/amlodipine: a review of its use in adult patients with essential hypertension not adequately controlled with monotherapy.
American journal of cardiovascular drugs : drugs, devices, and other interventions (2013-03-22)
Karly P Garnock-Jones
ABSTRACT
Combination therapy is often required in patients with hypertension, and fixed-dose single-pill combinations have been shown to provide an easier regimen for patients, improving adherence. Irbesartan/amlodipine (Aprovasc®) is an angiotensin-receptor blocker/calcium-channel blocker fixed-dose single-pill combination, whose constituent drugs exert additive effects when coadministered. In two randomized, open-label, multicentre, phase III trials, fixed-dose combination therapy with irbesartan/amlodipine was more effective than continuation of irbesartan or amlodipine monotherapy in patients with hypertension not adequately controlled with initial irbesartan or amlodipine monotherapy; there was a significantly greater decrease from baseline in mean seated home systolic blood pressure (primary endpoint) with the fixed-dose combination. The fixed-dose combination was also associated with a greater decrease in mean seated home diastolic blood pressure and mean seated office systolic and diastolic blood pressure than monotherapy. The fixed-dose combination of irbesartan/amlodipine was well tolerated in these patients; most treatment-emergent adverse events were of mild or moderate severity. The most frequent adverse event was peripheral oedema, generally associated with amlodipine treatment.
Sigma-Aldrich
Irbesartan, ≥98% (HPLC), powder
USP
Irbesartan, United States Pharmacopeia (USP) Reference Standard
Irbesartan, European Pharmacopoeia (EP) Reference Standard