Merck
STIM1 for stimulation of phagocyte NADPH oxidase.
Blood (2014-04-05)
Richard D Ye
ABSTRACT
In this issue of Blood, Zhang et al show that mice lacking the stromal-interacting molecule 1 (STIM1) gene in bone marrow cells are more susceptible to bacterial infection but are resistant to ischemia/reperfusion injury because of defective activation of phagocyte nicotinamide adenine dinucleotide phosphate (NADPH) oxidase.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard