Merck
Total synthesis of psoralidin, an anticancer natural product.
The Journal of organic chemistry (2009-03-04)
Pallab Pahari, Jürgen Rohr
ABSTRACT
A base-catalyzed condensation of phenyl acetate with acid chloride, followed by intramolecular cyclization and microwave-assisted cross-metathesis reaction, leads to the first total synthesis of psoralidin, a natural product with a broad range of biological activities, in a highly convergent and regioselective manner.
Sigma-Aldrich
Psoralidin, ≥98% (HPLC)