Merck
  • Enantioselective conversion of primary alcohols to α-exo-methylene γ-butyrolactones via iridium-catalyzed C-C bond-forming transfer hydrogenation: 2-(alkoxycarbonyl)allylation.
Enantioselective conversion of primary alcohols to α-exo-methylene γ-butyrolactones via iridium-catalyzed C-C bond-forming transfer hydrogenation: 2-(alkoxycarbonyl)allylation.
Journal of the American Chemical Society (2012-06-28)
T Patrick Montgomery, Abbas Hassan, Boyoung Y Park, Michael J Krische
ABSTRACT
Upon exposure of acrylic ester 1 to alcohols 2a-i in the presence of a cyclometalated iridium catalyst modified by (-)-TMBTP, catalytic C-C coupling occurs, providing enantiomerically enriched 5-substituted α-exo-methylene γ-butyrolactones 3a-i. Bromination of the methylene butyrolactone products followed by zinc-mediated reductive aldehyde addition provides the disubstituted α-exo-methylene γ-butyrolactones 6a and 6b with good to excellent levels of diastereoselectivity.
Sigma-Aldrich
α-Methylene-γ-butyrolactone, 97%