Merck
Photodegradation of folate sensitized by riboflavin.
Photochemistry and photobiology (2011-03-08)
Regina S Scurachio, Leif H Skibsted, Gustavo Metzker, Daniel R Cardoso
ABSTRACT
Folate is shown to react with singlet-excited state of riboflavin in a diffusion controlled reaction and with triplet-excited state of riboflavin in a somewhat slower reaction with (3)k(q) = 4.8 × 10(8) L mol(-1) s(-1) in aqueous phosphate buffer at pH 7.4, ionic strength of 0.2 mol L(-1), and 25°C. Singlet quenching is assigned as photo-induced reductive electron transfer from ground state folate to singlet-excited riboflavin, while triplet quenching is assigned as one-electron transfer rather than hydrogen atom transfer from folate to triplet-excited riboflavin, as the reaction quantum yield, φ = 0.32, is hardly influenced by solvent change from water to deuterium oxide, φ = 0.37. Cyclic voltammetry showed an irreversible two-electron anodic process for folate, E = 1.14 V versus NHE at a scan-rate of 50 mV s(-1), which appears to be kinetically controlled by the heterogeneous electron transfer from the substrates to the electrode. Main products of folate photooxidation sensitized by riboflavin were pterin-6-carboxylic acid and p-aminobenzoyl-L-glutamic acid as shown by liquid chromatographic ion-trap mass spectrometry (LC-IT-MS).
Sigma-Aldrich
N-(4-Aminobenzoyl)-L-glutamic acid, ≥98% (TLC)
Folic acid impurity A, European Pharmacopoeia (EP) Reference Standard