Merck
  • Impairment of sulfobromophthalein biliary excretion and inhibition of glutathione S-transferase activity induced by perhexiline maleate in rats.
Impairment of sulfobromophthalein biliary excretion and inhibition of glutathione S-transferase activity induced by perhexiline maleate in rats.
Biochemical pharmacology (1988-09-15)
M A Mariscal, M E Muñoz, P S Collado, A Esteller, J Gonzalez
ABSTRACT
The effect of the antianginal agent perhexiline maleate (160 mg/kg i.g., daily for 4 days) on the biliary excretion of sulfobromophthalein (BSP) and BSP-glutathione and the hepatic activity of glutathione S-transferases was investigated in Wistar rats. Perhexiline maleate caused a significant reduction in the maximal biliary excretion of BSP (-28%). The decrease corresponded to a lowered excretion of the conjugated dye whereas the excretion of the parent compound did not change significantly. Administration of the drug caused no effect on the maximal biliary excretion of infused BSP-glutathione. Liver glutathione concentrations were similar in control and treated rats. Perhexiline maleate significantly reduced liver glutathione S-transferase activities toward BSP (-25%), 3,4-dichloronitrobenzene (DCNB) (-21%) and 1-chloro-3,4-dinitrobenzene (DNCB) (-27%). Kinetic studies of the enzyme in liver cytosol showed that perhexiline maleate induced an uncompetitive inhibition for the BSP substrate with a reduced Vmax and Km. The results indicate that the reduction in glutathione S-transferase activity plays an important role as a factor determining the impairment in the hepatobiliary transport of BSP caused by perhexiline maleate.
Sigma-Aldrich
Perhexiline maleate salt, ≥98% (HPLC)