Merck
[Stability of PEGylated puerarin under different storage conditions].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-02-05)
Xin-Yi Liu, Da-Xiong Xiang, Jian-He Li, Xiong-Bin Hu, Zu-Guang Ye
ABSTRACT
To detect the stability of PEGylated puerarin (PEG-PUE), in order to provide experimental basis for storage conditions of PEGylated puerarin. First, a method for determining the content of PEG-PUE was established. Next, a system study was conducted for the stability of PEG-PUE affected by different factors such as temperature, humidity, light and light avoidance. PEG-PUE was severely degraded under the conditions of high temperature, high humidity and light. It was also seriously degraded under high temperature. PEG-PUE shall be stored under low temperature and in a dark and dry environment.
Sigma-Aldrich
Puerarin, ≥98.0% (HPLC)
Supelco
Puerarin, analytical standard