Merck
Structural and magnetic properties of Ba2LuMoO6: a valence bond glass.
Journal of physics. Condensed matter : an Institute of Physics journal (2013-01-25)
Fiona C Coomer, Edmund J Cussen
ABSTRACT
We report here the synthesis of the site ordered double perovskite Ba(2)LuMoO(6). Rietveld refinement of room temperature powder x-ray diffraction measurements indicates that it crystallizes in the cubic space group Fm3m, with a = 8.3265(1) Å. Powder neutron diffraction data indicate that, unusually, this cubic symmetry is maintained down to 2 K, with [Formula: see text], Mo(5+) ions situated on the frustrated face-centred cubic lattice. Despite dc-susceptibility measurements showing Curie-Weiss behaviour with strong antiferromagnetic interactions at T ≥ 200 K, there is no evidence of long range magnetic ordering at 2 K. At T ≤ 50 K, susceptibility measurements indicate a loss in moment to ∼18% of the expected value, and there is a corresponding loss in the magnitude of the magnetic exchange. The structural and magnetic properties of this compound are compared with the related compound Ba(2)YMoO(6), which is a valence bond glass.
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Sigma-Aldrich
Calcium titanate, nanopowder, <100 nm particle size (BET), 99% trace metals basis