Merck
  • Inhibition of prostaglandin biosynthesis during postluteolysis and effects on CL regression, prolactin, and ovulation in heifers.
Inhibition of prostaglandin biosynthesis during postluteolysis and effects on CL regression, prolactin, and ovulation in heifers.
Theriogenology (2012-05-15)
G Pugliesi, F A Khan, M A Hannan, M A Beg, G R Carvalho, O J Ginther
ABSTRACT
The beginning of postluteolysis (progesterone, <1 ng mL(-1)) in heifers was targeted by using 8 h after ultrasonic detection of a 25% decrease in CL area (cm2) and was designated Hour 0. Flunixin meglumine (FM; n=10) to inhibit PGF2α secretion or vehicle (n=9) were given intramuscularly at Hours 0, 4, 8, 16, 24, 32, and 40. The dose of FM was 2.5 mg/kg at each treatment. Blood sampling and measurement of the CL and dominant follicle were done every 8 h beginning 14 days postovulation in each group. Blood samples for detection of pulses of PRL and pulses of a metabolite of PGF2α (PGFM) were obtained every hour for 24 h beginning at Hour 0. Pulse concentrations of both PGFM and PRL were lower in the FM group than in the vehicle group. Concentration of PRL was greatest at the peak of a PGFM pulse. Neither CL area (cm2) nor progesterone concentration differed between groups during Hours 0 to 48 (postluteolysis). Ovulation occurred in nine of nine heifers in the vehicle group and in three of 10 heifers in the FM group. The anovulatory follicles in the FM group grew to 36.2±2.9 mm, and the wall became thickened from apparent luteinization. The hypothesis that PGF2α was involved in the continued P4 decrease and structural CL regression during postluteolysis was not supported. However, the hypotheses that pulses of PGFM and PRL were temporally related and that systemic FM treatment induced an anovulatory follicle were supported.
Sigma-Aldrich
Clonixin, ≥98% (HPLC)
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)