Merck
  • Rhodium-catalyzed carbonylation of 3-acyloxy-1,4-enynes for the synthesis of cyclopentenones.
Rhodium-catalyzed carbonylation of 3-acyloxy-1,4-enynes for the synthesis of cyclopentenones.
Organic letters (2012-03-03)
Xiaoxun Li, Suyu Huang, Casi M Schienebeck, Dongxu Shu, Weiping Tang
ABSTRACT
Functionalized cyclopentenones were synthesized by a Rh-catalyzed carbonylation of 3-acyloxy-1,4-enynes, derived from alkynes and α,β-unsaturated aldehydes. The reaction involved a Saucy-Marbet 1,3-acyloxy migration of propargyl esters and a [4 + 1] cycloaddition of the resulting acyloxy substituted vinylallene with CO.
Sigma-Aldrich
2-Cyclopenten-1-one, 98%