Merck
  • Production of 3-hydroxypropionic acid through propionaldehyde dehydrogenase PduP mediated biosynthetic pathway in Klebsiella pneumoniae.
Production of 3-hydroxypropionic acid through propionaldehyde dehydrogenase PduP mediated biosynthetic pathway in Klebsiella pneumoniae.
Bioresource technology (2011-10-25)
Lian Hua Luo, Chul Ho Kim, Sun-Yeon Heo, Baek-Rock Oh, Won-Kyung Hong, Seonghun Kim, Dae-Hyuk Kim, Jeong-Woo Seo
ABSTRACT
The pduP gene encodes a propionaldehyde dehydrogenase (PduP) was investigated for the role in 3-hydroxypropionic acid (3-HP) glycerol metabolism in Klebsiella pneumoniae. The enzyme assay showed that cell extracts from a pduP mutant strain lacked measurable dehydrogenase activity. Additionally, the mutant strain accumulated the cytotoxic intermediate metabolite 3-hydroxypropionaldehyde (3-HPA), causing both cell death and a lower final 3-HP titer. Ectopic expression of pduP restored normal cell growth to mutant. The enzymatic property of recombinant protein from Escherichia coli was examined, exhibiting a broad substrate specificity, being active on 3-HPA. The present work is thus the first to demonstrate the role of PduP in glycerol metabolism and biosynthesis of 3-HP.
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Propionaldehyde, 97%
Sigma-Aldrich
Propionaldehyde, reagent grade, 97%