Merck
  • The spectroscopic (FTIR, FT-IR gas phase and FT-Raman), first order hyperpolarizabilities, NMR analysis of 2,4-dichloroaniline by ab initio HF and density functional methods.
The spectroscopic (FTIR, FT-IR gas phase and FT-Raman), first order hyperpolarizabilities, NMR analysis of 2,4-dichloroaniline by ab initio HF and density functional methods.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-03-03)
N Sundaraganesan, J Karpagam, S Sebastian, J P Cornard
ABSTRACT
In this work, the experimental and theoretical study on molecular structure and vibrational spectra of 2,4-dichloroaniline (2,4-DCA) were studied. The Fourier transform infrared (gas phase) and Fourier transform Raman spectra of 2,4-DCA were recorded. The molecular geometry and vibrational frequencies of 2,4-DCA in the ground state were calculated by using the Hartree-Fock (HF) and density functional (DF) methods (BLYP, B3LYP and SVWN) with 6-31G(d,p) as basis set. Comparison of the observed fundamental vibrational frequencies of 2,4-DCA with calculated results by HF and density functional methods indicates that BLYP is superior to other methods for molecular vibrational problems. The difference between the observed and scaled wave number values of most of the fundamentals is very small. The electric dipole moment (micro) and the first hyperpolarizability (beta) values of the investigated molecule were computed using ab initio quantum mechanical calculations. The calculated results also show that the 2,4-DCA molecule might have microscopic nonlinear optical (NLO) behavior with non-zero values. Natural atomic charges of 2,4-DCA and 4-chloroaniline was calculated and compared. The isotropic chemical shift computed by (13)C NMR analyses also shows good agreement with experimental observations. The theoretically predicted FTIR and FT-Raman spectra of the title molecule have been constructed.
Sigma-Aldrich
2,4-Dichloroaniline, 99%
Supelco
2,4-Dichloroaniline, PESTANAL®, analytical standard