Merck
  • Phthalate esters (PAEs): emerging organic contaminants in agricultural soils in peri-urban areas around Guangzhou, China.
Phthalate esters (PAEs): emerging organic contaminants in agricultural soils in peri-urban areas around Guangzhou, China.
Environmental pollution (Barking, Essex : 1987) (2008-03-18)
Feng Zeng, Kunyan Cui, Zhiyong Xie, Lina Wu, Min Liu, Guoquan Sun, Yujun Lin, Danling Luo, Zunxiang Zeng
ABSTRACT
This study reports the first data on the concentrations and distribution of phthalate esters (PAEs) in the agricultural soils from the peri-urban areas of Guangzhou city. Sigma(16)PAEs concentrations ranged from 0.195 to 33.6 microg g(-1)-dry weight (dw). Elevated levels of PAEs were recorded in the vegetable fields located next to the urban districts, and a decreasing trend exists following the distance away from the urban center. Diisobutyl phthalate (DiBP), Di-n-butyl phthalate (DnBP), and Di(2-ethylhexyl) phthalate (DEHP) dominated the PAEs in the agricultural soils. Significant relationship (correlation coefficient R(2)=0.85, p<0.01, n=40) was present between the accumulation of PAEs and total organic carbons in agricultural soils. In addition, both pH and texture of soils are found to be important factors affecting the level of PAEs. This study shows that the agricultural soils in the peri-urban area of Guangzhou city were moderately polluted by PAEs.
Sigma-Aldrich
Diisobutyl phthalate, 99%
Supelco
Diisobutyl phthalate, certified reference material, TraceCERTĀ®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland