Merck
beta-carotene is converted primarily to retinoids in rats in vivo.
The Journal of nutrition (2000-08-05)
A B Barua, J A Olson
ABSTRACT
beta-Carotene might be converted oxidatively to vitamin A- active products in animals by the following three possible routes: 1) central cleavage, 2) sequential excentric cleavage or 3) random cleavage. Central cleavage is strongly favored by stoichiometric studies with tissue homogenates in vitro. To examine the relative importance of these pathways in rats in vivo, an oral dose (5.6 micromol) of all-trans beta-carotene in oil was given to vitamin A-deficient (-A) and to vitamin A-sufficient (+A) adult female Sprague-Dawley rats. Serum and several tissues were analyzed before and 3 h after dosing. The primary products of beta-carotene found in the intestine, serum and liver were retinol, retinyl esters and retinoic acid. Two minor oxidation products of beta-carotene, namely, 5,6-epoxy-beta-carotene and a partially characterized hydroxy-beta-carotene, were present in the stomach and its contents as well as in intestinal preparations. In the intestine, including its contents, of -A rats, very minor amounts of 5,6-epoxyretinyl palmitate and of beta-apocarotenals (8', 10', 12', 14') were identified. The total amount of the beta-apocarotenoids, however, was <5% of the retinoids formed in the intestine from beta-carotene during the same period. Another beta-carotene derivative, with a spectrum similar to that of semi-beta-carotenone, citranaxanthin and beta-apo-6'-carotenal, was also found in the intestinal extract of a -A rat. beta-Apocarotenals, beta-apocarotenols, beta-apocarotenyl esters and beta-apocarotenoic acids were not detected in tissues of +A rats nor in other tissues of -A rats. These findings agree with the view that central cleavage is by far the major pathway for the formation of vitamin A from beta-carotene in healthy rats in vivo.
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ≥96.0% (UV)
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ~20% apocarotenal basis (UV-vis), suspension (oily)