Merck
  • Determination of estrogens in human urine by high-performance liquid chromatography/diode array detection with ultrasound-assisted cloud-point extraction.
Determination of estrogens in human urine by high-performance liquid chromatography/diode array detection with ultrasound-assisted cloud-point extraction.
Analytical biochemistry (2011-11-01)
Yan Zou, Yanhua Li, Hua Jin, Huinan Tang, Deqing Zou, Mousheng Liu, Yaling Yang
ABSTRACT
A novel and efficient analytical methodology is proposed for extracting and preconcentrating three kinds of estrogens (17β-estradiol (βE2), estrone (E1), and diethylstilbestrol (DES)) in human urine prior to high-performance liquid chromatography (HPLC) analysis. It is based on the induction of micellar organized medium by using a nonionic surfactant (Tergitol TMN-6) to extract the target estrogens. Ultrasound was applied to enhance the extraction efficiency. Parameters affecting the extraction of target analytes including the concentration of surfactant, temperature, extraction time, sample pH, ionic strength, and centrifuging time were investigated. Under the optimum conditions, the linear range of βE2, E1, and DES was from 5.0 to 1000 ng/ml. All correlation coefficients of the calibration curves were higher than 0.997. The relative standard deviations (RSD, n=5) were 2.36-5.27% and the limits of detection (LOD) were 0.1, 0.2, and 0.1 ng/ml for βE2, E1, and DES in human urine, respectively. The results indicated that the method was successfully applied for analyzing βE2, E1, and DES in human urine.
Sigma-Aldrich
TERGITOL, Type 15-S-7
Pricing and availability is not currently available.
Sigma-Aldrich
TERGITOL TMN 10, Non-ionic, 1313 ppm, liquid, cloud point 76 °C
Pricing and availability is not currently available.