Merck
  • Distribution of androgen and oestrogen receptors-α in the seminal vesicle-related spinal neurones in male rats.
Distribution of androgen and oestrogen receptors-α in the seminal vesicle-related spinal neurones in male rats.
Journal of neuroendocrinology (2013-02-19)
X Q Sun, C Xu, P Leclerc, F Giuliano, G Benoît, S Droupy
ABSTRACT
The seminal vesicles are male accessory sex glands that contribute much of the seminal fluid volume. Previous studies have suggested that the majority of autonomic innervations to the rat seminal vesicles originate from the bilateral major pelvic ganglia. Many preganglionic autonomic neurones innervating the pelvic ganglion were expressed androgen receptors (AR) or oestrogen receptor (ER)-α immunoreactivity. However, direct neuroanatomic data regarding the distribution of AR and ER-α in seminal vesicle related-spinal neurones are lacking. In the present study, a nonvirulent pseudorabies virus (PRV-152 strain) was used in a retrograde tracing experiment. Four days after PRV injection into the seminal vesicles of male rats, spinal cord sections were prepared. Double- and triple-fluorescence techniques using AR and ER-α with choline acetyltransferase (ChAT) and PRV were used to investigate the AR and ER-α distribution in the seminal vesicles related spinal neurones in male rats. In lamina X, 14% of the PRV-labelled neurones in the L1-L4 segments and 43% in the L5-S1 segments were double-labelled with AR. In the L1-L4 segments, 6% of PRV-labelled neurones and 26% in the L5-S1 segments were double-labelled with ER-α. In the intermedial cell column area, 10% of PRV-labelled neurones in the L1-L4 segments and 47% of PRV-labelled neurones in the L5-S1 segments were double-labelled with AR. Up to 16% of PRV-labelled neurones in the L5-S1 segments were double-labelled with ER-α. No PRV-labelled neurones in the L1-L4 segments were double-labelled with ER-α. However, for the AR and ER-α/PRV/ChAT triple-fluorescence experiments, very few seminal vesicle preganglionic neurones expressed AR or ER-α. Our data suggests that many spinal interneurones but not preganglionic neurones involved in the seminal vesicle control in male rats were double-labelled with AR or ER-α, and they were mainly located at the parasympathetic level in the spinal cord.
Sigma-Aldrich
Anti-Androgen Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution