Merck
  • Structure-based design of novel quinoxaline-2-carboxylic acids and analogues as Pim-1 inhibitors.
Structure-based design of novel quinoxaline-2-carboxylic acids and analogues as Pim-1 inhibitors.
European journal of medicinal chemistry (2018-05-21)
Bruno Oyallon, Marie Brachet-Botineau, Cédric Logé, Pascal Bonnet, Mohamed Souab, Thomas Robert, Sandrine Ruchaud, Stéphane Bach, Pascal Berthelot, Fabrice Gouilleux, Marie-Claude Viaud-Massuard, Caroline Denevault-Sabourin
ABSTRACT
We identified a new series of quinoxaline-2-carboxylic acid derivatives, targeting the human proviral integration site for Moloney murine leukemia virus-1 (HsPim-1) kinase. Seventeen analogues were synthesized providing useful insight into structure-activity relationships studied. Docking studies realized in the ATP pocket of HsPim-1 are consistent with an unclassical binding mode of these inhibitors. The lead compound 1 was able to block HsPim-1 enzymatic activity at nanomolar concentrations (IC50 of 74 nM), with a good selectivity profile against a panel of mammalian protein kinases. In vitro studies on the human chronic myeloid leukemia cell line KU812 showed an antitumor activity at micromolar concentrations. As a result, compound 1 represents a promising lead for the design of novel anticancer targeted therapies.
Sigma-Aldrich
Histone from calf thymus, Type III-S
Sigma-Aldrich
Indirubin-3′-oxime, ≥98% (HPLC), solid
Sigma-Aldrich
Staurosporine from Streptomyces sp., for molecular biology, ≥95% (HPLC)
Sigma-Aldrich
CHR-6494 trifluoroacetate salt, ≥98% (HPLC)