Merck
  • Dissociation of chick embryonic tissue for FACS and preparation of isolated cells for genome-wide downstream assays.
Dissociation of chick embryonic tissue for FACS and preparation of isolated cells for genome-wide downstream assays.
STAR protocols (2021-04-20)
Ruth M Williams, Tatjana Sauka-Spengler
ABSTRACT
In order to process samples by fluorescence-activated cell sorting (FACS), it is essential to obtain a single-cell suspension of dissociated cells. Numerous protocols and commercial reagents are available; however, each requires optimization for specific tissue types. Here, we describe an optimized protocol for dissociating dissected chick embryos across a broad span of developmental stages. We also provide protocols for processing targeted cell populations isolated using FACS for ATAC-seq, RNA-seq, and chromatin immunoprecipitation. For complete details on the use and execution of this protocol, please refer to Ling and Sauka-Spengler (2019) and Williams et al. (2019).
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Pricing and availability is not currently available.