Merck
  • Thick ascending limb claudins are altered to increase calciuria and magnesiuria in metabolic acidosis.
Thick ascending limb claudins are altered to increase calciuria and magnesiuria in metabolic acidosis.
American journal of physiology. Renal physiology (2021-02-02)
Il Hwan Oh, Chor Ho Jo, Sua Kim, Sungsin Jo, Sungjin Chung, Gheun-Ho Kim
ABSTRACT
Urinary calcium and magnesium wasting is a characteristic feature of metabolic acidosis, and this study focused on the role of the thick ascending limb of Henle's loop in metabolic acidosis-induced hypercalciuria and hypermagnesiuria because thick ascending limb is an important site of paracellular calcium and magnesium reabsorption. Male Sprague-Dawley rats were used to determine the effects of acid loading (by adding NH4Cl, 7.2 mmol/220 g body wt/day to food slurry for 7 days) on renal expression of claudins and then to evaluate whether the results were reversed by antagonizing calcium-sensing receptor (using NPS-2143). At the end of each animal experiment, the kidneys were harvested for immunoblotting, immunofluorescence microscopy, and quantitative PCR (qPCR) analysis of claudins and the calcium-sensing receptor. As expected, NH4Cl loading lowered urinary pH and increased excretion of urinary calcium and magnesium. In NH4Cl-loaded rats, renal protein and mRNA expression of claudin-16, and claudin-19, were decreased compared with controls. However, claudin-14 protein and mRNA increased in NH4Cl-loaded rats. Consistently, the calcium-sensing receptor protein and mRNA were up-regulated in NH4Cl-loaded rats. All these changes were reversed by NPS-2143 coadministration and were confirmed using immunofluorescence microscopy. Hypercalciuria and hypermagnesiuria in NH4Cl-loaded rats were significantly ameliorated by NPS-2143 coadministration as well. We conclude that in metabolic acidosis, claudin-16 and claudin-19 in the thick ascending limb are down-regulated to produce hypercalciuria and hypermagnesiuria via the calcium-sensing receptor.NEW & NOTEWORTHY This study found that the thick ascending limb of Henle's loop is involved in the mechanisms of hypercalciuria and hypermagnesiuria in metabolic acidosis. Specifically, expression of claudin-16/19 and claudin-14 was altered via up-regulation of calcium-sensing receptor in NH4Cl-induced metabolic acidosis. Our novel findings contribute to understanding the regulatory role of paracellular tight junction proteins in the thick ascending limb.
Sigma-Aldrich
Cinacalcet hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O