Merck
  • Serological comparison of systemic lupus erythematosus with neuropsychiatric lupus using synthetic nucleic acid antigens.
Serological comparison of systemic lupus erythematosus with neuropsychiatric lupus using synthetic nucleic acid antigens.
Journal of translational autoimmunity (2020-11-19)
Sangita Khatri, Nikos Psaraftis, Alessia Funaro, Yoshiyuki Arinuma, Yuichiro Fujieda, Simone Mader, Christian Damsgaard Jørgensen, Kira Astakhova
ABSTRACT
Neuropsychiatric systemic lupus erythematosus is an autoimmune disorder characterized by an irregular exchange between the central nervous system and the immune system, leading to the outbreak of neurological conditions with possible disabling effects. Although neuropsychiatric systemic lupus erythematosus is the most common expression of lupus condition, it is still poorly understood. In this study, we focus on the development of an advantageous method based on the application of synthetic nucleic acids and protein-based antigen arrays in order to characterize autoreactive antibodies in neuropsychiatric systemic lupus erythematosus. We confirmed the benefits of using synthetic oligonucleotides such as assay reproducibility, elevated affinity and specificity to autoreactive antibodies. We also demonstrated presence of autoantibodies towards three particular synthetic double stranded antigens and verify similarity of antinuclear antibody patterns in ordinary lupus and neuropsychiatric systemic lupus erythematosus.
Sigma-Aldrich
Anti-GP120 antibody produced in rabbit, affinity isolated antibody