Merck
  • High-phosphorus diets reduce aortic lesions and cardiomyocyte size and modify lipid metabolism in Ldl receptor knockout mice.
High-phosphorus diets reduce aortic lesions and cardiomyocyte size and modify lipid metabolism in Ldl receptor knockout mice.
Scientific reports (2020-11-29)
Sarah M Grundmann, Alexandra Schutkowski, Christian Berger, Anja C Baur, Bettina König, Gabriele I Stangl
ABSTRACT
The consumption of phosphorus in Western populations largely exceeds the recommended intake, while vitamin D supply is often insufficient. Both situations are linked to an increased cardiovascular risk. A 17-week two-factorial study with Ldl receptor-/- mice was conducted to investigate the cardiovascular impact of dietary phosphorus [adequate (0.3%; P0.3) vs. high (1.5%; P1.5)] in combination with a low (50 IU/kg; D50) or adequate vitamin D diet (1000 IU/kg; D1000). The data demonstrate that mice fed the P1.5 vs. P0.3 diets developed smaller vascular lesions (p = 0.013) and cardiac hypotrophy (p = 0.011), which were accompanied by diminished IGF1 and insulin signalling activity in their hearts. Vitamin D showed no independent effect on atherogenesis and heart morphology. Feeding P1.5 vs. P0.3 diets resulted in markedly reduced serum triacylglycerols (p < 0.0001) and cholesterol (p < 0.0001), higher faecal lipid excretion (p < 0.0001) and a reduced mRNA abundance of hepatic sterol exporters and lipoprotein receptors. Minor hypocholesterolaemic and hypotriglyceridaemic effects were also found in mice fed the D1000 vs. D50 diets (p = 0.048, p = 0.026). To conclude, a high phosphorus intake strongly affected the formation of vascular lesions, cardiac morphology, and lipid metabolism, although these changes are not indicative of an increased cardiovascular risk.
Sigma-Aldrich
Cholesteryl oleate, ≥98% (HPLC; detection at 205 nm)
Sigma-Aldrich
cis-10-Heptadecenoic acid, ≥99%