Merck
  • Nonlinearities between inhibition and T-type calcium channel activity bidirectionally regulate thalamic oscillations.
Nonlinearities between inhibition and T-type calcium channel activity bidirectionally regulate thalamic oscillations.
eLife (2020-09-10)
Adam C Lu, Christine Kyuyoung Lee, Max Kleiman-Weiner, Brian Truong, Megan Wang, John R Huguenard, Mark P Beenhakker
ABSTRACT
Absence seizures result from 3 to 5 Hz generalized thalamocortical oscillations that depend on highly regulated inhibitory neurotransmission in the thalamus. Efficient reuptake of the inhibitory neurotransmitter GABA is essential, and reuptake failure worsens human seizures. Here, we show that blocking GABA transporters (GATs) in acute rat brain slices containing key parts of the thalamocortical seizure network modulates epileptiform activity. As expected, we found that blocking either GAT1 or GAT3 prolonged oscillations. However, blocking both GATs unexpectedly suppressed oscillations. Integrating experimental observations into single-neuron and network-level computational models shows how a non-linear dependence of T-type calcium channel gating on GABAB receptor activity regulates network oscillations. Receptor activity that is either too brief or too protracted fails to sufficiently open T-type channels necessary for sustaining oscillations. Only within a narrow range does prolonging GABAB receptor activity promote channel opening and intensify oscillations. These results have implications for therapeutics that modulate inhibition kinetics.
Sigma-Aldrich
(S)-SNAP-5114, ≥98% (HPLC), solid
Sigma-Aldrich
NO-711 hydrochloride, ≥98% (HPLC)