Merck
  • Statins exacerbate glucose intolerance and hyperglycemia in a high sucrose fed rodent model.
Statins exacerbate glucose intolerance and hyperglycemia in a high sucrose fed rodent model.
Scientific reports (2019-06-21)
Sriram Seshadri, Naimisha Rapaka, Bhumika Prajapati, Dipeeka Mandaliya, Sweta Patel, Christopher Shamir Muggalla, Bandish Kapadia, Phanithi Prakash Babu, Parimal Misra, Uday Saxena
ABSTRACT
Statins are first-line therapy drugs for cholesterol lowering. While they are highly effective at lowering cholesterol, they have propensity to induce hyperglycemia in patients. Only limited studies have been reported which studied the impact of statins on (a) whether they can worsen glucose tolerance in a high sucrose fed animal model and (b) if so, what could be the molecular mechanism. We designed studies using high sucrose fed animals to explore the above questions. The high sucrose fed animals were treated with atorvastatin and simvastatin, the two most prescribed statins. We examined the effects of statins on hyperglycemia, glucose tolerance, fatty acid accumulation and insulin signaling. We found that chronic treatment with atorvastatin made the animals hyperglycemic and glucose intolerant in comparison with diet alone. Treatment with both statins lead to fatty acid accumulation and inhibition of insulin signaling in the muscle tissue at multiple points in the pathway.
Sigma-Aldrich
Anti-phospho-IRS1 (Ser307 mouse/ Ser312 human) Antibody, clone 24.6.2, clone 24.6.2, from mouse