
Z-Triprolidine hydrochloride

Synonym(s):
2-[(1Z)-1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propen-1-yl]pyridine monohydrochloride, (Z)-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride
Empirical Formula (Hill Notation):
C19H22N2 · HCl
CAS Number:
Molecular Weight:
314.85
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing