
Trizma® hydrochloride solution

Synonym(s):
Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride
Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Products
Brand
Formula Weight
pH Value
pK Value
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing