
Tranylcypromine Related Compound B

Synonym(s):
trans-Tranylcypromine hydrochloride, (2E)-3-Phenyl-2-propen-1-amine hydrochloride, trans-Cinnamylamine hydrochloride
Empirical Formula (Hill Notation):
C9H11N · HCl
CAS Number:
Molecular Weight:
169.65
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing