
Rifamycin Sodium

Synonym(s):
Rifamycin SV sodium salt
Empirical Formula (Hill Notation):
C37H46NNaO12
CAS Number:
Molecular Weight:
719.75
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2