
Pyrimidifen

Synonym(s):
5-Chloro-N-{2-[4-(2-ethoxyethyl)-2,3-dimethylphenoxy]ethyl}-6-ethylpyrimidin-4-amine
Empirical Formula (Hill Notation):
C20H28ClN3O2
CAS Number:
Molecular Weight:
377.91
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2