
PTAD-Azide

Synonym(s):
4-(4-(2-Azidoethoxy)phenyl)-1,2,4-triazolidine-3,5-dione, N3-Ph-Ur for e-Y-CLICK
Empirical Formula (Hill Notation):
C10H10N6O3
CAS Number:
Molecular Weight:
262.22
Products
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing